Learn More
Abcam 4-Trifluoromethylumbelliferyl-ß-D-galactopyranoside (TFMU-Gal), 25MG
Supplier:  Abcam AB27507025MG
Encompass_Preferred
MW 392.28 Da, Purity >95%. Blue-green fluorogenic probe with higher fluorescence in the physiological pH range due to lower pKa. Suitable for the detection of LacZ without the need of a stop buffer treatment step for development of fluorescence.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-996-089