Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sigma Aldrich Trifluoromethanesulfonic Acid
Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Description
Trifluoromethanesulfonic acid is a strong organic acid. It can be prepared by reacting bis(trifluoromethylthio)mercury with H2O2. On mixing with HNO3, it affords a nitrating reagent (a nitronium salt). This reagent is useful for the nitration of aromatic compounds. Its dissociation in various organic solvents has been studied.Trifluoromethanesulfonic acid can be used as a catalyst to prepare: Substituted tetrahydrofurans and tetrahydropyrans by cyclization of corresponding unsaturated alcohols under acidic conditions. Nitriles from corresponding aldehydes by Schmidt reaction. Disubstituted five-membered ring lactones by allylboration reaction between 2-alkoxycarbonyl allylboronates and aldehydes. It can also be used as a catalyst in the Fischer glycosylation and Friedel-Crafts acylation reactions.ReagentPlus is a registered trademark of Sigma-Aldrich Co. LLC
Specifications
Specifications
| CAS | 1493-13-6 |
| Density | 1.696 g/mL at 25°C |
| Boiling Point | 162°C |
| Molecular Formula | CHF3O3S |
| Refractive Index | n20/D 1.327 |
| Linear Formula | CF3SO3H |
| MDL Number | MFCD00007514 |
| Quantity | 5 g |
| Synonym | Triflic acid |
| Molecular Weight (g/mol) | 150.08 g/mol |
| Show More |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction