Learn More
Strem, An Ascensus Company CAS# 1493-13-6. 10g. Trifluoromethanesulfonic acid, 99+%. MFCD00007514
SDP
Supplier:  Strem, An Ascensus Company 09741010G
Encompass
CAS# 1493-13-6. 10g. Trifluoromethanesulfonic acid, 99+%. MFCD00007514. Molecular Weight: 150.07. Molecular Formula: CF3SO3H. Color/form: colorless to light brown fuming liq. Strem# 09-7410. http://www.strem.com/catalog/v/09-7410/
Catalog No. 50-901-09437