Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Trifluoromethanesulfonic Acid

Catalog No. 347817100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-781-7100G 100 g
34-781-725G 25 g
34-781-7500G 500 g
34-781-75G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Danger
Catalog No. 34-781-7100G Supplier Sigma Aldrich Supplier No. 347817100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Trifluoromethanesulfonic acid is a strong organic acid. It can be prepared by reacting bis(trifluoromethylthio)mercury with H2O2. On mixing with HNO3, it affords a nitrating reagent (a nitronium salt). This reagent is useful for the nitration of aromatic compounds. Its dissociation in various organic solvents has been studied.Trifluoromethanesulfonic acid can be used as a catalyst to prepare: Substituted tetrahydrofurans and tetrahydropyrans by cyclization of corresponding unsaturated alcohols under acidic conditions. Nitriles from corresponding aldehydes by Schmidt reaction. Disubstituted five-membered ring lactones by allylboration reaction between 2-alkoxycarbonyl allylboronates and aldehydes. It can also be used as a catalyst in the Fischer glycosylation and Friedel-Crafts acylation reactions.ReagentPlus is a registered trademark of Sigma-Aldrich Co. LLC

Specifications

CAS 1493-13-6
Density 1.696 g/mL at 25°C
Boiling Point 162°C
Molecular Formula CHF3O3S
Refractive Index n20/D 1.327
Linear Formula CF3SO3H
MDL Number MFCD00007514
Quantity 100 g
Synonym Triflic acid
Molecular Weight (g/mol) 150.08 g/mol
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.