Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trifluoromethanesulfonic acid solution, Solstice

Catalog No. 6001763 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-017-63 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-017-63 Supplier Solstice Supplier No. 353171L

missing translation for 'validMainframeMsg'

Volumetric, 0.1 M CF3SO3H in acetic acid (0.1N), for titration in non-aqueous medium

Solstice's high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use
  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1493-13-6
Molecular Formula CHF3O3S
Molecular Weight (g/mol) 150.07
MDL Number MFCD00007514
InChI Key ITMCEJHCFYSIIV-UHFFFAOYSA-N
Synonym triflic acid, perfluoromethanesulfonic acid, trifluoromethanesulphonic acid, tfmsa, trifluoromethane sulfonic acid, tfoh, methanesulfonic acid, trifluoro, hotf, trifluoromethylsulfonic acid, trimsylate
PubChem CID 62406
ChEBI CHEBI:48511
IUPAC Name trifluoromethanesulfonic acid
SMILES OS(=O)(=O)C(F)(F)F

Specifications

Flash Point 38°C (100.4°F)
Quantity 1 L
Linear Formula CF3SO3H
UN Number UN2789
Beilstein 3693487
Formula Weight 150.08g/mol
Chemical Name or Material Trifluoromethanesulfonic acid solution
Hazard Statement P280-P305 + P351 + P338-P310
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.