Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Trifluoromethanesulfonic Anhydride Solution

Catalog No. 70408325ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-408-325ML 25 mL
70-408-3100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 70-408-325ML Supplier Sigma Aldrich Supplier No. 70408325ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Trifluoromethanesulfonic anhydride solution is generally used to convert hydroxyl groups to triflic esters. It can be used to prepare triflic azide for azido amino acid synthesis.

Specifications

CAS 358-23-6
Density 1.359 g/mL at 25°C
Molecular Formula C2F6O5S2
Refractive Index n20/D 1.402
Linear Formula C2F6O5S2
MDL Number MFCD00000408
Quantity 25 mL
Synonym Perfluoromethanesulfonic anhydride solution; Triflic anhydride solution
Molecular Weight (g/mol) 282.14 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.