Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trifluoromethanesulfonyl chloride, 99%

Catalog No. p-4510484
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC173050050 5 g Glass bottle
AC173050250 25 g Glass bottle
AC173051000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. AC173050050 Supplier Thermo Scientific Chemicals Supplier No. 173050050
Only null left
Add to Cart
This item is not returnable. View return policy

CAS: 421-83-0 | CClF3O2S | 168.51 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 421-83-0
Molecular Formula CClF3O2S
Molecular Weight (g/mol) 168.51
MDL Number MFCD00007451
InChI Key GRGCWBWNLSTIEN-UHFFFAOYSA-N
Synonym trifluoromethanesulphonyl chloride, triflyl chloride, methanesulfonyl chloride, trifluoro, trifluoromethanesulfonylchloride, trifluoro-methanesulfonyl chloride, methanesulfonylchloride, trifluoro, trifluoromethylsulfonyl chloride, triflic chloride, pfc-msc, cf3so2cl
PubChem CID 79000
IUPAC Name trifluoromethanesulfonyl chloride
SMILES FC(F)(F)S(Cl)(=O)=O

Specifications

Color Colorless
Density 1.5800g/mL
Boiling Point 29.0°C to 32.0°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.3330 to 1.3350
Linear Formula CF3SO2Cl
Quantity 5 g
Beilstein 03,IV,35
Fieser 09,485
Specific Gravity 1.58
Solubility Information Solubility in water: reacts
Formula Weight 168.52
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Trifluoromethanesulfonyl chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 207-009-

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.