Learn More
Strem, An Ascensus Company CAS# 358-23-6. 25g. Trifluoromethanesulphonic anhydride, min. 97%. MFCD00000408
SDP
Catalog No. 5090109440
Encompass
CAS# 358-23-6. 25g. Trifluoromethanesulphonic anhydride, min. 97%. MFCD00000408. Molecular Weight: 282.13. Molecular Formula: (CF3SO2)2O. Color/form: colorless liq. Strem# 09-7420. http://www.strem.com/catalog/v/09-7420/
Catalog No. 50-901-09440 Supplier Strem, An Ascensus Company Supplier No. 09742025G
CAS# 358-23-6. 25g. Trifluoromethanesulphonic anhydride, min. 97%. MFCD00000408. Molecular Weight: 282.13. Molecular Formula: (CF3SO2)2O. Color/form: colorless liq. Strem# 09-7420. http://www.strem.com/catalog/v/09-7420/