Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(Trifluoromethyl)trimethylsilane, 95%

Catalog No. AAB2034714
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2034714 25 g
AAB2034703 1 g
AAB2034706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2034714 Supplier Thermo Scientific Chemicals Supplier No. B2034714
  • $689.00
    $550.65 / Each of 1

    Save $138.35

    20% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 81290-20-2 | C5H11F3Si | 156.22 g/mol

It is employed as a valuable reagent for trifluoromethylation of electrophilic substrates. It is also use an the nucleophilic addition of the trifluoromethyl group to aldehydes and ketones. It generates trifluoromethide with catalytic amounts of F- (TBAF or CsF) and reacts with carbonyl compounds to produce trifluoromethylated alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a valuable reagent for trifluoromethylation of electrophilic substrates. It is also use an the nucleophilic addition of the trifluoromethyl group to aldehydes and ketones. It generates trifluoromethide with catalytic amounts of F- (TBAF or CsF) and reacts with carbonyl compounds to produce trifluoromethylated alcohols.

Solubility
Soluble in alcohol, THF, ether, dichloromethane, aromatic and aliphatic hydrocarbons. Soluble in water(with white smokes).

Notes
Moisture sensitive. Store away from oxidizing agents and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81290-20-2
Molecular Formula C5H11F3Si
Molecular Weight (g/mol) 156.22
MDL Number MFCD00145454
InChI Key MTYSDPUZDZURPP-UHFFFAOYSA-N
Synonym trifluoromethyl trimethylsilane, trimethyl trifluoromethyl silane, trifluoromethyltrimethylsilane, ruppert's reagent, tms-cf3, tfmtms, silane, trimethyl trifluoromethyl, unii-a009786qpj, trimethylsilyl trifluoromethane, ruppert-prakash reagent
PubChem CID 552549
IUPAC Name trimethyl(trifluoromethyl)silane
SMILES CC(C)(C)[SiH2]C(F)(F)F

Specifications

Boiling Point 54°C to 55°C
Quantity 25 g
Flash Point −10°C (14°F)
Refractive Index 1.3305
Sensitivity Moisture sensitive
Density 0.962
Linear Formula (CH3)3SiCF3
UN Number UN1993
Beilstein 4241868
Solubility Information Soluble in alcohol,THF,ether,dichloromethane,aromatic and aliphatic hydrocarbons. Soluble in water(with white smokes).
Formula Weight 142.2
Percent Purity 98%
Chemical Name or Material (Trifluoromethyl)trimethylsilane
Show More Show Less
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.