Learn More
Medchemexpress LLC Trifluoromethyl-tubercidin | 1854086-05-7 | 99.1% | 334.25 g/mol | C12H13F3N4O4 | 10 MG
SDP
Catalog No. 5000288022
Encompass_Preferred
Trifluoromethyl-tubercidin is a nucleoside analog inhibitor of 2'-O-ribose methyltransferase 1 (MTr1). Supplied as a research-grade compound, it is used in antiviral and methyltransferase inhibition studies and is suitable for in vitro biochemical and cellular assays.
  • High reported purity: 99.12%.
  • Characterized by molecular formula C12H13F3N4O4 and molecular weight 334.25 g/mol.
  • Useful for antiviral research and studies of RNA methyltransferase activity.
  • Available in milligram pack sizes suitable for laboratory assays.
  • Provides a defined nucleoside analog tool for mechanistic and screening experiments.
Catalog No. 50-002-88022 Supplier Medchemexpress LLC Supplier No. HY15604810MG
Trifluoromethyl-tubercidin is a nucleoside analog inhibitor of 2'-O-ribose methyltransferase 1 (MTr1). Supplied as a research-grade compound, it is used in antiviral and methyltransferase inhibition studies and is suitable for in vitro biochemical and cellular assays.
  • High reported purity: 99.12%.
  • Characterized by molecular formula C12H13F3N4O4 and molecular weight 334.25 g/mol.
  • Useful for antiviral research and studies of RNA methyltransferase activity.
  • Available in milligram pack sizes suitable for laboratory assays.
  • Provides a defined nucleoside analog tool for mechanistic and screening experiments.