Learn More
Strem, An Ascensus Company CAS# 81290-20-2. 5g. Trifluoromethyltrimethylsilane, min. 97%. MFCD00145454
SDP
Supplier:  Strem, An Ascensus Company 1479905G
Encompass
CAS# 81290-20-2. 5g. Trifluoromethyltrimethylsilane, min. 97%. MFCD00145454. Molecular Weight: 142.20. Molecular Formula: CF3Si(CH3)3. Color/form: colorless liq. Strem# 14-7990. http://www.strem.com/catalog/v/14-7990/
Catalog No. 50-901-09629