Learn More
Cayman Chemical Trifluorothymidine, Purity: ≥98%, CAS: 70-00-8, 100 mg
SDP
Catalog No. 502980894
Encompass_Preferred
A derivative of thymidine (Item No. 20519) that exhibits antiviral and anticancer activity by inhibiting thymidylate synthase, thereby blocking nucleic acid synthesis, and by incorporating into DNA, which leads to DNA damage and apoptosis
Catalog No. 50-298-0894 Supplier Cayman Chemical Supplier No. 21366100 MG
A derivative of thymidine (Item No. 20519) that exhibits antiviral and anticancer activity by inhibiting thymidylate synthase, thereby blocking nucleic acid synthesis, and by incorporating into DNA, which leads to DNA damage and apoptosis