Learn More
Apexbio Technology LLC Triflupromazine-HCl
SDP
Catalog No. 5000569366
Encompass_Preferred
Triflupromazine hydrochloride (CAS No 1098-60-8) is a phenothiazine derivative used in neuropharmacological research due to its antipsychotic and antiemetic properties It originates from the broader class of phenothiazines which function primarily by blocking dopamine receptors particularly the D2 subtype thereby influencing neurotransmitter pathways involved in mood and behavior regulation In vitro its activity is observed in the nanomolar to low micromolar range demonstrating effective inhibition of dopamine-mediated signaling Researchers often employ this compound in cell models and animal studies to explore its potential in treating psychiatric disorders and its efficacy in drug screening Experimental concentrations typically depend on the specific research design ensuring adaptability across various study parameters
Catalog No. 50-005-69366 Supplier Apexbio Technology LLC Supplier No. C7040100
Triflupromazine hydrochloride (CAS No 1098-60-8) is a phenothiazine derivative used in neuropharmacological research due to its antipsychotic and antiemetic properties It originates from the broader class of phenothiazines which function primarily by blocking dopamine receptors particularly the D2 subtype thereby influencing neurotransmitter pathways involved in mood and behavior regulation In vitro its activity is observed in the nanomolar to low micromolar range demonstrating effective inhibition of dopamine-mediated signaling Researchers often employ this compound in cell models and animal studies to explore its potential in treating psychiatric disorders and its efficacy in drug screening Experimental concentrations typically depend on the specific research design ensuring adaptability across various study parameters