Learn More
Cayman Chemical Trifluralin, Purity: ≥98%, CAS: 1582-09-8, 100 mg
SDP
Catalog No. 502909818
Encompass_Preferred
A dinitroaniline herbicide; inhibits photosynthetic electron transport, ATP synthesis, and cytochrome f reduction in isolated spinach chloroplasts; decreases cotton (G. hirsutum) plant height when applied to the soil prior to planting at 1.68 kg/ha; induces ROS production in V79 fibroblasts and genotoxicity in isolated human peripheral lymphocytes from 2.5-500 µM; has been found as a contaminant in freshwater fish
Catalog No. 50-290-9818 Supplier Cayman Chemical Supplier No. 35476100 MG
A dinitroaniline herbicide; inhibits photosynthetic electron transport, ATP synthesis, and cytochrome f reduction in isolated spinach chloroplasts; decreases cotton (G. hirsutum) plant height when applied to the soil prior to planting at 1.68 kg/ha; induces ROS production in V79 fibroblasts and genotoxicity in isolated human peripheral lymphocytes from 2.5-500 µM; has been found as a contaminant in freshwater fish