Learn More
Research Products International Corp TRIFLURALIN, 250MG
SDP
Supplier:  Research Products International Corp T500200.25
Encompass_Preferred
Disrupts mitosis by inhibiting the formation of microtubules.
Catalog No. 50-213-699