Learn More
TARGETMOL CHEMICALS INC Trifluridine 200MG
SDP
Catalog No. 502876004
Encompass_Preferred
Also available in 1 mL, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Trifluridine (NSC-75520) is a fluorinated thymidine analog with potential antineoplastic activity. Trifluridine is incorporated into DNA and inhibits thymidylate synthase, resulting in inhibition of DNA synthesis, inhibition of protein synthesis, and apoptosis. This agent also exhibits antiviral activity. Purity 99.39%
Catalog No. 50-287-6004 Supplier TARGETMOL CHEMICALS INC Supplier No. T1428200MG
Also available in 1 mL, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Trifluridine (NSC-75520) is a fluorinated thymidine analog with potential antineoplastic activity. Trifluridine is incorporated into DNA and inhibits thymidylate synthase, resulting in inhibition of DNA synthesis, inhibition of protein synthesis, and apoptosis. This agent also exhibits antiviral activity. Purity 99.39%