Learn More
Selleck Chemical LLC Trifluridine (NSC 75520) S1778-1g
SDP
Catalog No. 503110768
Encompass_Preferred
Trifluridine (NSC 529182 NSC 75520 Viroptic Trifluorothymidine TFT) is an anti-herpesvirus antiviral agent by interacting viral DNA replication used primarily on the eye
Catalog No. 50-311-0768 Supplier Selleck Chemical LLC Supplier No. S17781G
Add to Cart
Add to Cart
Trifluridine (NSC 529182 NSC 75520 Viroptic Trifluorothymidine TFT) is an anti-herpesvirus antiviral agent by interacting viral DNA replication used primarily on the eye