Learn More
Selleck Chemical LLC Trifluridine (NSC 75520) S1778-50mg
SDP
Catalog No. 503110825
Encompass_Preferred
Trifluridine (NSC 529182 NSC 75520 Viroptic Trifluorothymidine TFT) is an anti-herpesvirus antiviral agent by interacting viral DNA replication used primarily on the eye
Catalog No. 50-311-0825 Supplier Selleck Chemical LLC Supplier No. S177850MG
Trifluridine (NSC 529182 NSC 75520 Viroptic Trifluorothymidine TFT) is an anti-herpesvirus antiviral agent by interacting viral DNA replication used primarily on the eye