Learn More
Medchemexpress LLC Trifluridine/tipiracil hydrochloride mixture | 733030-01-8 | 100.0% | 435.76 | 25 MG
SDP
Catalog No. 5000393059
Encompass_Preferred
Trifluridine/tipiracil hydrochloride mixture (TAS-102) is a potent and orally active nucleoside antitumor agent. It is composed of a 1:0.5 mixture (on a molar basis) of alpha,alpha,alpha-tri-fluorothymidine (FTD) and thymidine phosphorylase inhibitor (TPI). The antitumor activity of TAS-102 primarily stems from its inhibition of thymidylate synthase (TS) and its incorporation into DNA.
  • Prevents body weight loss in colorectal and gastric cancer mouse models.
  • Reduces relative tumor volume in colorectal and gastric cancer mouse models.
Catalog No. 50-003-93059 Supplier Medchemexpress LLC Supplier No. 25MGHY16478
Trifluridine/tipiracil hydrochloride mixture (TAS-102) is a potent and orally active nucleoside antitumor agent. It is composed of a 1:0.5 mixture (on a molar basis) of alpha,alpha,alpha-tri-fluorothymidine (FTD) and thymidine phosphorylase inhibitor (TPI). The antitumor activity of TAS-102 primarily stems from its inhibition of thymidylate synthase (TS) and its incorporation into DNA.
  • Prevents body weight loss in colorectal and gastric cancer mouse models.
  • Reduces relative tumor volume in colorectal and gastric cancer mouse models.