Learn More
Medchemexpress LLC Trifluridine/tipiracil hydrochloride mixture | 733030-01-8 | 100.0% | 435.76 | 50 MG
SDP
Catalog No. 5000391748
Encompass_Preferred
Trifluridine/tipiracil hydrochloride mixture (TAS-102) is a potent and orally active nucleoside antitumor agent. It is composed of a 1:0.5 molar mixture of alpha,alpha,alpha-tri-fluorothymidine (FTD) and thymidine phosphorylase inhibitor (TPI). The antitumor activity primarily stems from its inhibition of thymidylate synthase (TS) and its incorporation into DNA.
  • Potent and orally active nucleoside antitumor agent
  • Composed of a 1:0.5 molar mixture of alpha,alpha,alpha-tri-fluorothymidine (FTD) and thymidine phosphorylase inhibitor (TPI)
  • Inhibits thymidylate synthase (TS)
  • Incorporates into DNA for antitumor activity
Catalog No. 50-003-91748 Supplier Medchemexpress LLC Supplier No. HY1647850MG
Trifluridine/tipiracil hydrochloride mixture (TAS-102) is a potent and orally active nucleoside antitumor agent. It is composed of a 1:0.5 molar mixture of alpha,alpha,alpha-tri-fluorothymidine (FTD) and thymidine phosphorylase inhibitor (TPI). The antitumor activity primarily stems from its inhibition of thymidylate synthase (TS) and its incorporation into DNA.
  • Potent and orally active nucleoside antitumor agent
  • Composed of a 1:0.5 molar mixture of alpha,alpha,alpha-tri-fluorothymidine (FTD) and thymidine phosphorylase inhibitor (TPI)
  • Inhibits thymidylate synthase (TS)
  • Incorporates into DNA for antitumor activity