Learn More
Medchemexpress LLC Trifluridine/tipiracil hydrochloride mixture | 733030-01-8 | 99.98% | 435.76 | C10H11F3N2O5.1/2C9H11ClN4O2.1/2HCl | 10 MG
SDP
Catalog No. 5000289229
Encompass_Preferred
Trifluridine/tipiracil hydrochloride mixture is a 1:0.5 molar combination of trifluridine (a thymidine analog) and tipiracil (a thymidine phosphorylase inhibitor) used in oncology research to evaluate antitumor activity. It acts by inhibiting thymidylate synthase and incorporating trifluridine into DNA, making it useful for preclinical studies of nucleoside-based therapeutics.
  • Antitumor nucleoside mixture for preclinical research.
  • Dual mechanism: thymidylate synthase inhibition and DNA incorporation.
  • High purity suitable for analytical and biological assays.
  • Available in small research quantities, including 10 mg, and larger packs.
  • Batch-specific certificate of analysis provided, including HPLC purity.
Catalog No. 50-002-89229 Supplier Medchemexpress LLC Supplier No. HY1647810MG
Trifluridine/tipiracil hydrochloride mixture is a 1:0.5 molar combination of trifluridine (a thymidine analog) and tipiracil (a thymidine phosphorylase inhibitor) used in oncology research to evaluate antitumor activity. It acts by inhibiting thymidylate synthase and incorporating trifluridine into DNA, making it useful for preclinical studies of nucleoside-based therapeutics.
  • Antitumor nucleoside mixture for preclinical research.
  • Dual mechanism: thymidylate synthase inhibition and DNA incorporation.
  • High purity suitable for analytical and biological assays.
  • Available in small research quantities, including 10 mg, and larger packs.
  • Batch-specific certificate of analysis provided, including HPLC purity.