Learn More
Medchemexpress LLC Trifluridine (Trifluorothymidine) | 70-00-8 | 99.98% | C10H11F3N2O5 | 100 MG
SDP
Catalog No. 5000432122
Encompass_Preferred
Trifluridine is an irreversible and orally active thymidylate synthase inhibitor that suppresses DNA synthesis. It is an antiviral molecule and an anticancer agent, used in research for various infections and tumors.
Catalog No. 50-004-32122 Supplier Medchemexpress LLC Supplier No. HYA0061100MG
Trifluridine is an irreversible and orally active thymidylate synthase inhibitor that suppresses DNA synthesis. It is an antiviral molecule and an anticancer agent, used in research for various infections and tumors.