Learn More
Apexbio Technology LLC Triflusal 322-79-2 5mg
SDP
Catalog No. 502240462
Encompass_Preferred
Triflusal (CAS 322-79-2) is a small molecule inhibitor targeting cyclooxygenase-1 (COX-1) in platelets It acts by acetylating COX-1 leading to irreversible suppression of thromboxane B2 synthesis thereby modulating platelet aggregation This mechanism makes Triflusal a useful tool in investigating platelet function and thromboxane-mediated pathways in biomedical research with applications in studies of hemostasis thrombosis and antiplatelet drug development
This item is not returnable. View return policy
Catalog No. 50-224-0462 Supplier Apexbio Technology LLC Supplier No. B14615
Triflusal (CAS 322-79-2) is a small molecule inhibitor targeting cyclooxygenase-1 (COX-1) in platelets It acts by acetylating COX-1 leading to irreversible suppression of thromboxane B2 synthesis thereby modulating platelet aggregation This mechanism makes Triflusal a useful tool in investigating platelet function and thromboxane-mediated pathways in biomedical research with applications in studies of hemostasis thrombosis and antiplatelet drug development