Learn More
Cayman Chemical Trifolirhizin, Purity: ≥98%, CAS: 6807-83-6, 10 mg
SDP
Catalog No. 502950028
Encompass_Preferred
A flavonoid with anticancer and vasorelaxant activities; induces the accumulation of LC3 puncta in HCT116 and SW620 cells, as well as inhibits proliferation of B16/F10 murine melanoma and human A2780 ovarian and MKN45 gastric cancer cells at 20 µM; induces intratumor apoptosis and necrosis and reduces tumor weight and volume in a colorectal cancer mouse xenograft model; induces relaxation in precontracted isolated mouse aortic rings in a β2-adrenergic receptor-independent manner from 0.2-20 µg/ml
Catalog No. 50-295-0028 Supplier Cayman Chemical Supplier No. 3465510 MG
A flavonoid with anticancer and vasorelaxant activities; induces the accumulation of LC3 puncta in HCT116 and SW620 cells, as well as inhibits proliferation of B16/F10 murine melanoma and human A2780 ovarian and MKN45 gastric cancer cells at 20 µM; induces intratumor apoptosis and necrosis and reduces tumor weight and volume in a colorectal cancer mouse xenograft model; induces relaxation in precontracted isolated mouse aortic rings in a β2-adrenergic receptor-independent manner from 0.2-20 µg/ml