Learn More
Medchemexpress LLC TriGalNAc CBz | 186613-57-0 | 98.7% | 633.64 | C34H35NO11 | 10 MG
SDP
Catalog No. 5000287105
Encompass_Preferred
TriGalNAc CBz is a GalNAc derivative used in research to target the asialoglycoprotein receptor (ASGPR) for hepatocyte-directed delivery. It is applied in mRNA delivery and lysosome-targeting chimera (LYTAC) studies, and serves as a chemical linker for GalNAc-LYTAC synthesis.
  • Acts as an ASGPR ligand for hepatocyte targeting.
  • Applicable for mRNA drug delivery research.
  • Enables synthesis of lysosome-targeting chimeras (LYTACs) as a linker.
  • High purity (98.7%) suitable for research applications.
  • Solid white to off-white appearance for easy handling.
  • Available as solid and as a DMSO solution for flexible use.
  • Defined storage conditions for long-term stability.
Catalog No. 50-002-87105 Supplier Medchemexpress LLC Supplier No. HY14735410MG
TriGalNAc CBz is a GalNAc derivative used in research to target the asialoglycoprotein receptor (ASGPR) for hepatocyte-directed delivery. It is applied in mRNA delivery and lysosome-targeting chimera (LYTAC) studies, and serves as a chemical linker for GalNAc-LYTAC synthesis.
  • Acts as an ASGPR ligand for hepatocyte targeting.
  • Applicable for mRNA drug delivery research.
  • Enables synthesis of lysosome-targeting chimeras (LYTACs) as a linker.
  • High purity (98.7%) suitable for research applications.
  • Solid white to off-white appearance for easy handling.
  • Available as solid and as a DMSO solution for flexible use.
  • Defined storage conditions for long-term stability.