Learn More
MSC Rust-Oleum Trigger Spray Bottle, 32 oz, Plastic, Mist/Stream, 1 ea
Catalog No. 502867193
Encompass
This empty spray bottle with a trigger is designed for a variety of liquids including cleaners and pesticides. It features a durable plastic construction and offers both mist and stream dispensing types.
  • Plastic bottle and nozzle construction
  • Red and clear trigger sprayer for easy identification
  • Features an 11-inch dip tube length
  • Plastic seal material ensures leak-resistant operation
  • Graduated bottle for precise liquid measurement
  • Overall fractional diameter of 5
  • Suitable for cleaners, pesticides, and various liquids
  • Dispensing options include mist and stream
Catalog No. 50-286-7193 Supplier MSC Supplier No. 02605301
This empty spray bottle with a trigger is designed for a variety of liquids including cleaners and pesticides. It features a durable plastic construction and offers both mist and stream dispensing types.
  • Plastic bottle and nozzle construction
  • Red and clear trigger sprayer for easy identification
  • Features an 11-inch dip tube length
  • Plastic seal material ensures leak-resistant operation
  • Graduated bottle for precise liquid measurement
  • Overall fractional diameter of 5
  • Suitable for cleaners, pesticides, and various liquids
  • Dispensing options include mist and stream