Learn More
MSC BluShield Pressure Washer Gun, Brass, 3000 PSI, Metric
Catalog No. 502835906
Encompass
This pressure washer gun is a precision-engineered tool designed for metric pressure washing tasks. It ensures seamless pairing with metric pressure washers and accessories, offering precision, power, and adaptability for cleaning tasks.
  • Precision-engineered for metric pressure washers.
  • Compatible with metric wands and accessories.
  • Male metric inlet accommodates hoses with female metric fittings for a secure connection.
  • Female metric outlet directly connects to metric wands, enhancing setup efficiency.
  • Equipped with a trigger lock to prevent accidental spraying and ensure controlled operation.
Catalog No. 50-283-5906 Supplier MSC Supplier No. 63959761
This pressure washer gun is a precision-engineered tool designed for metric pressure washing tasks. It ensures seamless pairing with metric pressure washers and accessories, offering precision, power, and adaptability for cleaning tasks.
  • Precision-engineered for metric pressure washers.
  • Compatible with metric wands and accessories.
  • Male metric inlet accommodates hoses with female metric fittings for a secure connection.
  • Female metric outlet directly connects to metric wands, enhancing setup efficiency.
  • Equipped with a trigger lock to prevent accidental spraying and ensure controlled operation.