Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ Trigger Sprayer Pumps

Catalog No. 02991563
Change view
Click to view available options
Liner Material:
F422 Foam Polyethylene
No. per Case:
144 Cs.
Packaging:
Bagged
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Liner Material No. per Case Packaging
02-991-563 F422 Foam Polyethylene 144 Cs. Bagged
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-991-563 Supplier Qorpak™ Supplier No. CAP06051

missing translation for 'validMainframeMsg'

Dispense liquid from nozzle which allows user to adjust spray pattern from jet stream to spray to fine mist

  • Made of white polypropylene
  • Available with PE Foam liner or industrial sprayers with strainer,stainless-steel balls and springs

Pump with F217 Foam PE liner

  • For general use trigger spray bottles
Unlined heavy-duty pump with F217 gasket
  • For industrial chemical trigger spray bottles
  • Chemical resistant

Specifications

Color White
Material Polypropylene
No. per Case 144 Cs.
No. per Pack 12/Pk.
Packaging Bagged
Sterility Non-sterile
Liner Material F422 Foam Polyethylene
Neck Finish 28-400
For Use With (Equipment) Trigger spray bottle
Autoclavable Not Autoclavable
Disposable Reusable
Product Type Threaded Screw Cap
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.