Learn More
Apexbio Technology LLC TRIGLYCIDYL ISOCYANURATE-1g
SDP
Catalog No. 5000897574
Encompass_Preferred
Triglycidyl Isocyanurate (Teroxirone) (CAS No 2451-62-9) is a compound commonly used in biomedical research due to its ability to modify proteins and nucleic acids Originating from synthetic chemistry it primarily acts by alkylating nucleophilic sites in biological molecules interfering with DNA and protein functions Its mechanism involves the formation of cross-links which is a crucial factor in its in vitro activity often in the nanomolar to low micromolar range This compound is utilized in cell models and animal studies to investigate cellular responses and for drug screening applications Experimental concentrations typically depend on the specific research design
Catalog No. 50-008-97574 Supplier Apexbio Technology LLC Supplier No. C70921000
Triglycidyl Isocyanurate (Teroxirone) (CAS No 2451-62-9) is a compound commonly used in biomedical research due to its ability to modify proteins and nucleic acids Originating from synthetic chemistry it primarily acts by alkylating nucleophilic sites in biological molecules interfering with DNA and protein functions Its mechanism involves the formation of cross-links which is a crucial factor in its in vitro activity often in the nanomolar to low micromolar range This compound is utilized in cell models and animal studies to investigate cellular responses and for drug screening applications Experimental concentrations typically depend on the specific research design