Learn More
Medchemexpress LLC Triglycine | 556-33-2 | MFCD00036223 | 99.7% | 189.17 g/mol | C6H11N3O4 | 1 G
SDP
Catalog No. 5000302648
Encompass_Preferred
H-Gly-Gly-Gly-OH (Triglycine) is a simple tripeptide made of three glycine residues. Supplied as a white to off-white solid, it is used as a model compound, reference standard, and reagent in peptide synthesis, biochemical assays, and protein studies.
  • Simple tripeptide composed of three glycine residues.
  • High purity suitable for analytical and biochemical applications.
  • White to off-white solid, convenient for handling and storage.
  • Stable under recommended storage conditions for reliable results.
  • Useful as a reference standard and model compound in peptide chemistry and protein assays.
Catalog No. 50-003-02648 Supplier Medchemexpress LLC Supplier No. HYW0152361G
H-Gly-Gly-Gly-OH (Triglycine) is a simple tripeptide made of three glycine residues. Supplied as a white to off-white solid, it is used as a model compound, reference standard, and reagent in peptide synthesis, biochemical assays, and protein studies.
  • Simple tripeptide composed of three glycine residues.
  • High purity suitable for analytical and biochemical applications.
  • White to off-white solid, convenient for handling and storage.
  • Stable under recommended storage conditions for reliable results.
  • Useful as a reference standard and model compound in peptide chemistry and protein assays.