Learn More
Medchemexpress LLC Trigonelline | 535-83-1 | MFCD00054262 | 100.0% | 137.14 g/mol | C7H7NO2 | 10 MG
SDP
Catalog No. 5000295980
Encompass_Preferred
Trigonelline is a naturally occurring alkaloid used as a research reagent and analytical standard. It functions as an Nrf2 inhibitor and has reported antidiabetic, antibacterial, antifungal, and antiviral activities, and can induce ferroptosis. Supplied as a white to light yellow solid, it is intended for biological assays and analytical applications.
  • High purity suitable for analytical and biological studies.
  • Well-characterized molecular weight and chemical formula.
  • Available in small milligram quantities for research use.
  • Solid form; store protected from light at recommended temperatures.
  • Appropriate as a reference standard for Nrf2 and apoptosis research.
Catalog No. 50-002-95980 Supplier Medchemexpress LLC Supplier No. HYN041410MG
Trigonelline is a naturally occurring alkaloid used as a research reagent and analytical standard. It functions as an Nrf2 inhibitor and has reported antidiabetic, antibacterial, antifungal, and antiviral activities, and can induce ferroptosis. Supplied as a white to light yellow solid, it is intended for biological assays and analytical applications.
  • High purity suitable for analytical and biological studies.
  • Well-characterized molecular weight and chemical formula.
  • Available in small milligram quantities for research use.
  • Solid form; store protected from light at recommended temperatures.
  • Appropriate as a reference standard for Nrf2 and apoptosis research.