Learn More
Medchemexpress LLC Trigonelline | 535-83-1 | MFCD00054262 | 137.14 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYN041450MG
Encompass_Preferred
Trigonelline is an alkaloid with potential antidiabetic activity. It acts as a potent Nrf2 inhibitor, blocking Nrf2-dependent proteasome activity and enhancing apoptosis in pancreatic cancer cells. It also exhibits anti-HSV-1, antibacterial, and antifungal activities, and induces ferroptosis.
  • Potential antidiabetic activity
  • Potent Nrf2 inhibitor
  • Enhances apoptosis in pancreatic cancer cells
  • Antimicrobial properties (anti-HSV-1, antibacterial, antifungal)
  • Induces ferroptosis
  • Cardioprotective activity by rescuing H9c2 cell morphology and attenuating H2O2 induced cell deaths
Catalog No. 50-003-88713