Learn More
Medchemexpress LLC HY-N0415 100mg Medchemexpress, Trigonelline chloride CAS:6138-41-6 Purity:>98%
SDP
Catalog No. 501933560
Encompass_Preferred
Medchemexpress, HY-N0415 100mg Trigonelline chloride CAS:6138-41-6 Trigonelline chloride, an alkaloid with potential antidiabetic activity, is present in considerable amounts in coffee. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3560 Supplier Medchemexpress LLC Supplier No. HYN0415100MG
Medchemexpress, HY-N0415 100mg Trigonelline chloride CAS:6138-41-6 Trigonelline chloride, an alkaloid with potential antidiabetic activity, is present in considerable amounts in coffee. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.