Learn More
Medchemexpress LLC Trigonelline-d3 chloride | 6138-41-6 | 99.6% | 176.62 | C7H5D3ClNO2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN0415S10MG
Encompass_Preferred
Trigonelline-d3 chloride is a deuterium-labeled analytical standard of trigonelline hydrochloride supplied as a white to off-white solid intended for research use. It provides a stable-isotope internal standard for quantitative GC- or LC-MS assays.
  • Deuterium-labeled trigonelline chloride for use as an internal standard.
  • High purity (99.6%) suitable for analytical applications.
  • White to off-white solid; solid form for easy handling and storage.
  • Soluble in DMSO (5 mg/mL) with warming or sonication recommended.
  • Stable under sealed storage at 4°C; shorter-term storage guidance for solutions provided.
  • Available in small pack sizes (including 10 mg) for standard laboratory use.
Catalog No. 50-002-96376