Learn More
Description
Specifications
Specifications
| CAS | 6138-41-6 |
| Physical Form | Neat |
| Linear Formula | C7H7 NO2 · HCl |
| MDL Number | MFCD00077250 |
| Synonym | 1-Methylpyridinium-3-carboxylate hydrochloride; N-Methylnicotinic acid betaine hydrochloride |
| Type | Analytical Standard |
| Quantity | 5 g |
| Formula Weight | 173.60 |
| Grade | Analytical Standard |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.