Learn More
Cayman Chemical Dioscin, Purity: ≥95%, CAS: 19057-60-4, 10 mg
SDP
Catalog No. 502951644
Encompass_Preferred
A natural steroid saponin that inhibits proliferation and induces apoptosis in cancer cells; has antioxidant activity and induces autophagy
Catalog No. 50-295-1644 Supplier Cayman Chemical Supplier No. 1183410 MG
A natural steroid saponin that inhibits proliferation and induces apoptosis in cancer cells; has antioxidant activity and induces autophagy