Learn More
Cayman Chemical Trigonelline-d3 (chloride), Purity: ≥99% deuterated forms (d1-d3), 10 mg
SDP
Catalog No. 502912171
Encompass_Preferred
An internal standard for the quantification of trigonelline by GC- or LC-MS
Catalog No. 50-291-2171 Supplier Cayman Chemical Supplier No. 3634010 MG
An internal standard for the quantification of trigonelline by GC- or LC-MS