Learn More
Med Vet International Merck, Tri-Hart Plus Green, 26-50Lb, 136mcg, Chew Tablets, 60 Dose
Catalog No. 502828337
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Prevents heartworm disease in dogs and puppies 6 weeks and older by eliminating the tissue stage of heartworm larvae (Dirofilaria immitis) for 30 days after infection; treats and controls ascarids (T. canis, T. leonina) and hookworms (A. caninum. U. stenocephala, A. braziliense). Active ingredients: ivermectin and pyrantel. Once-a-month dosing.
Catalog No. 50-282-8337 Supplier Med Vet International Supplier No. RXTRIHARTGR
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Prevents heartworm disease in dogs and puppies 6 weeks and older by eliminating the tissue stage of heartworm larvae (Dirofilaria immitis) for 30 days after infection; treats and controls ascarids (T. canis, T. leonina) and hookworms (A. caninum. U. stenocephala, A. braziliense). Active ingredients: ivermectin and pyrantel. Once-a-month dosing.