Learn More
Medchemexpress LLC Triheptanoin (UX007) | 620-67-7 | 99.9% | 428.61 | 100 MG
SDP
Catalog No. 5000454358
Encompass_Preferred
Triheptanoin (Propane-1,2,3-triyl triheptanoate) is a synthetic medium-chain triglyceride (MCT) consisting of three odd-chain 7-carbon (heptanoate) fatty acids on a glycerol backbone. It can be used for the research of inherited metabolic disorders.
  • Used as an anaplerotic compound in long-chain fatty acid oxidation disorders (LC-FAOD), providing calories and fatty acids to bypass enzyme deficiencies.
  • Can be used for research of ataxia-telangiectasia (A-T).
  • Functions as an anaplerotic agent replenishing tricarboxylic acid (TCA) cycle intermediates by metabolism to heptanoate (C7), acetyl-CoA, and propionyl-CoA, supplying energy.
  • Mitigates brain ATP depletion and mitochondrial dysfunction in a mouse model of Alzheimer's disease.
  • Preserved mitochondrial functions in the hippocampal formations of pilocarpine-induced status epilepticus (SE) mice.
Catalog No. 50-004-54358 Supplier Medchemexpress LLC Supplier No. HYW013136100MG
Triheptanoin (Propane-1,2,3-triyl triheptanoate) is a synthetic medium-chain triglyceride (MCT) consisting of three odd-chain 7-carbon (heptanoate) fatty acids on a glycerol backbone. It can be used for the research of inherited metabolic disorders.
  • Used as an anaplerotic compound in long-chain fatty acid oxidation disorders (LC-FAOD), providing calories and fatty acids to bypass enzyme deficiencies.
  • Can be used for research of ataxia-telangiectasia (A-T).
  • Functions as an anaplerotic agent replenishing tricarboxylic acid (TCA) cycle intermediates by metabolism to heptanoate (C7), acetyl-CoA, and propionyl-CoA, supplying energy.
  • Mitigates brain ATP depletion and mitochondrial dysfunction in a mouse model of Alzheimer's disease.
  • Preserved mitochondrial functions in the hippocampal formations of pilocarpine-induced status epilepticus (SE) mice.