Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Trihexyl phosphate | 2528-39-4 | MFCD00015351 | >90.0% | 350.47 g·mol⁻1 | C18H39O4P | 10 MG
SDP

Catalog No. 5000281947
Encompass_Preferred

Trihexyl phosphate is an organophosphate research reagent commonly used as a flame retardant additive and as a test compound in in-vitro toxicology and material studies. It has reported effects on mitochondrial function and estrogenic activity in cell models. Intended for laboratory research use only, recommended storage is at room temperature for long-term stability or in solvent at low temperatures for extended preservation.

  • Organophosphate compound used as a flame retardant and research reagent.
  • CAS number 2528-39-4.
  • Molecular formula C18H39O4P and molecular weight 350.47 g·mol⁻1.
  • Available in small milligram pack sizes suitable for laboratory use.
  • Storage: room temperature (stable 3 years); in solvent store at -80°C (2 years) or -20°C (1 year).
  • For research use only; not for human or clinical use.

Catalog No. 50-002-81947 Supplier Medchemexpress LLC Supplier No. HYW28848010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Trihexyl phosphate is an organophosphate research reagent commonly used as a flame retardant additive and as a test compound in in-vitro toxicology and material studies. It has reported effects on mitochondrial function and estrogenic activity in cell models. Intended for laboratory research use only, recommended storage is at room temperature for long-term stability or in solvent at low temperatures for extended preservation.

  • Organophosphate compound used as a flame retardant and research reagent.
  • CAS number 2528-39-4.
  • Molecular formula C18H39O4P and molecular weight 350.47 g·mol⁻1.
  • Available in small milligram pack sizes suitable for laboratory use.
  • Storage: room temperature (stable 3 years); in solvent store at -80°C (2 years) or -20°C (1 year).
  • For research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.