Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Trihexyl phosphate | 2528-39-4 | MFCD00015351 | >90.0% | 350.47 g/mol | C18H39O4P | 25 MG
SDP

Catalog No. 5000280768
Encompass_Preferred

Trihexyl phosphate (CAS 2528-39-4) is an organophosphate compound used as a flame retardant and as a chemical reagent in laboratory research. It is supplied in small, milligram-scale quantities for analytical and in vitro work and requires standard chemical handling and storage precautions.

  • Organophosphate flame retardant and reagent.
  • Suitable for small-scale laboratory use and analytical testing.
  • Molecular formula C18H39O4P; molecular weight 350.47 g/mol.
  • CAS number 2528-39-4 for unambiguous identification.
  • Storage guidance: solid at room temperature; in solvent store at -80°C or -20°C per supplier documentation.
  • Reported biological effects in cell assays (mitochondrial impairment, apoptosis); use appropriate safety controls.

Catalog No. 50-002-80768 Supplier Medchemexpress LLC Supplier No. HYW28848025MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Trihexyl phosphate (CAS 2528-39-4) is an organophosphate compound used as a flame retardant and as a chemical reagent in laboratory research. It is supplied in small, milligram-scale quantities for analytical and in vitro work and requires standard chemical handling and storage precautions.

  • Organophosphate flame retardant and reagent.
  • Suitable for small-scale laboratory use and analytical testing.
  • Molecular formula C18H39O4P; molecular weight 350.47 g/mol.
  • CAS number 2528-39-4 for unambiguous identification.
  • Storage guidance: solid at room temperature; in solvent store at -80°C or -20°C per supplier documentation.
  • Reported biological effects in cell assays (mitochondrial impairment, apoptosis); use appropriate safety controls.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.