Learn More
Medchemexpress LLC Trihexyl phosphate | 2528-39-4 | MFCD00015351 | 98.0% | 350.47 g/mol | C18H39O4P | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW28848050MG
Encompass_Preferred
Trihexyl phosphate is an organophosphate compound used as a flame retardant and reagent in material science and laboratory research. It is a hydrophobic, low-volatility liquid suitable for testing, formulation, and small-scale experiments.
  • Organophosphate flame retardant and plasticizer for polymers and coatings.
  • Liquid, colorless to light yellow, hydrophobic.
  • Low volatility and suitable for formulation studies and material testing.
  • Commonly used in small research quantities for experimental work.
  • CAS number 2528-39-4; molecular weight ~350.47 g/mol.
Catalog No. 50-002-82164