Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trihexyl(tetradecyl)phosphonium Dicyanamide 93.0+%, TCI America™
SDP

Catalog No. T35345G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T35345G 5 g
T353425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T35345G Supplier TCI America Supplier No. T35345G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 701921-71-3
Molecular Formula C34H68N3P
Molecular Weight (g/mol) 549.91
MDL Number MFCD04117699
InChI Key DOMOOBQQQGXLGU-UHFFFAOYSA-N
PubChem CID 16217238
IUPAC Name [(cyanoimino)methylidene]azanide; trihexyl(tetradecyl)phosphanium
SMILES [N-]=C=NC#N.CCCCCCCCCCCCCC[P+](CCCCCC)(CCCCCC)CCCCCC

Specifications

Quantity 5 g
UN Number 3265
Formula Weight 549.91
Percent Purity ≥93.0% (N)
Physical Form Liquid
Chemical Name or Material Trihexyl(tetradecyl)phosphonium Dicyanamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.