Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Trihexyltetradecylphosphonium Bis(2,4,4-Trimethylpentyl)Phosphinate

Catalog No. 286125GF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
28-612-5GF 5 g
28-612-50GF 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 28-612-5GF Supplier Sigma Aldrich Supplier No. 286125GF
Only null left
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

It can be used as an extractant for the removal of dibenzothiophene fro mLiquid fuels via extractive desulfurization (EDS).A phosphonium phosphinate for use as a polar solvent

Specifications

CAS 465527-59-7
Density 0.895 g/mL at 20°C
Molecular Formula C48H102O2P2
Linear Formula C48H102O2P2
MDL Number MFCD04117696
Quantity 5 g
Synonym Tetradecyltrihexylphosphonium bis(2,4,4-trimethylpentyl)phosphinate
Molecular Weight (g/mol) 773.27 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.