Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Trihexyltetradecylphosphonium Bis(Trifluoromethylsulfonyl)Amide

Catalog No. 509715GF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-971-5GF 5 g
50-971-50GF 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 50-971-5GF Supplier Sigma Aldrich Supplier No. 509715GF
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Trihexyltetradecylphosphonium bis(trifluoromethylsulfonyl)amide [P6,6,6,14][Ntf2] is an ionic liquid generally used in biosensors electrodeposition, and synthetic chemistry applications.[P6,6,6,14][Ntf2] can be used in: The electrodeposition of nano and microcrystalline metals for electrochemical applications. The synthesis of amino acid ionic liquid functionalized graphene (AAIL-GR) applicable as a biosensor. The investigation of interactions between ionic liquids and fluorinated alkanes. The synthesis of pure rutile and anatase phases of nanocrystalline TiO2.Ionic liquid for copolymerisation

Specifications

CAS 460092-03-9
Density 1.07 g/mL at 20°C
Molecular Formula C34H68F6NO4PS2
Linear Formula [CH3(CH2)5]3P[N(SO2CF3)2](CH2)13CH3
MDL Number MFCD04117695
Quantity 5 g
Synonym Tetradecyltrihexylphosphonium bis(trifluoromethylsulfonyl)amide
Molecular Weight (g/mol) 764 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.