Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Trihexyltetradecylphosphonium Chloride

Catalog No. 8974450GF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-744-50GF 5 g
89-744-5GF 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 89-744-50GF Supplier Sigma Aldrich Supplier No. 8974450GF
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Trihexyltetradecylphosphonium chloride is a hydrophobic ionic liquid, which is soluble in carbon dioxide.Trihexyltetradecylphosphonium chloride can be used as binder to develop a silver particle-modified carbon paste electrode, which can determine nitrite ions at low concentration.

Specifications

CAS 153086-78-3
Density 1.046 g/mL at 20°C
Molecular Formula C11H24N2O4
Linear Formula C11H24N2O4
MDL Number MFCD03788155
Quantity 5 g
Synonym Tetradecyltrihexylphosphonium chloride
Molecular Weight (g/mol) 248.32 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.