Learn More
Research Products International Corp 2,3,5-Triiodobenzoic Acid, 10 Grams
SDP
Supplier:  Research Products International Corp T5004010.0
Encompass_Preferred
Non-competitive inhibitor of polar auxin transport.
Catalog No. 50-213-700