Learn More
Selleck Chemical LLC Triiodothyronine 25mg 6893-02-3 T3
SDP
Supplier:  Selleck Chemical LLC S572625MG
Encompass_Preferred
Triiodothyronine (T3) is a thyroid hormone that affects various physiological process in the body, including growth and development, metabolism, body temperature, and heart rate.This product is soluble but easy to precipitate after freezing in DMSO. NaOH is recommended as a stock solution. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8243