Learn More
MOLECULAR DEPOT LLC Triiodothyronine BSA - 5 mg
Catalog No. 5000013489
Encompass_Preferred
Triiodothyronine BSA Conjugate is a concentrated solution of Triiodothyronine (T3) conjugated to BSA A carbon spacer arm links BSA to the Thyroxine moiety Each BSA molecule contains several Triiodothyronine units Dilutions of the provided conjugate can be made in Phosphate Buffer Saline pH 7 4 For in vitro research use only Not for human or animal use
Catalog No. 50-000-13489 Supplier MOLECULAR DEPOT LLC Supplier No. B2012115
Triiodothyronine BSA Conjugate is a concentrated solution of Triiodothyronine (T3) conjugated to BSA A carbon spacer arm links BSA to the Thyroxine moiety Each BSA molecule contains several Triiodothyronine units Dilutions of the provided conjugate can be made in Phosphate Buffer Saline pH 7 4 For in vitro research use only Not for human or animal use