Learn More
Revvity Health Sciences Inc Triiodothyronine, L-3, 5, 3'-[125I]-, 500 µCi (18.5MBq), Specific Activity: 1080-1320 µCi (40-48.8 MBq)/µg
Catalog No. 502810994
Encompass
Triiodothyronine, L-3, 5, 3'-[125I]-, 500 µCi (18.5MBq), Specific Activity: 1080-1320 µCi (40-48.8 MBq)/µg
Catalog No. 50-281-0994 Supplier Revvity Health Sciences Inc Supplier No. NEX110H500UC
Triiodothyronine, L-3, 5, 3'-[125I]-, 500 µCi (18.5MBq), Specific Activity: 1080-1320 µCi (40-48.8 MBq)/µg